Please login to your online account to display your discounted pricing

Mouse anti-Complement C4b/d, Clone: SPM545, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437405

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Antigen Complement C4b/d
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant human Complement 4d protein
Isotype IgG1, κ
Regulatory Status RUO
Gene ID (Entrez) 721
Reconstitution Protein A purified
Clone SPM545
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Test Specificity This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.
Concentration 0.2mg/mL