Please login to your online account to display your discounted pricing

Mouse anti-Complement C4b/d, Clone: SPM545, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23437401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23437401

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant human Complement 4d protein
Conjugate Unlabeled
Antigen Complement C4b/d
Format Purified
Formulation PBS with 0.05% BSA.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 721
Test Specificity This MAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d.
Reconstitution Protein A purified
Clone SPM545
Concentration 0.2mg/mL
Cross Reactivity Human