Please login to your online account to display your discounted pricing

Mouse anti-Common beta Chain, Clone: 1C1, Novus Biologicals™

$219.00 - $219.00

Specifications

Antigen Common beta Chain
Clone 1C1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD131.
Gene ID (Entrez) 1439
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14335403

 
novus biologicals canada
NBP1433540025MG
0.025mg Each for $219.00
Description & Specifications

Specifications

Antigen Common beta Chain
Clone 1C1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD131.
Gene ID (Entrez) 1439
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting