Please login to your online account to display your discounted pricing

Mouse anti-Common beta Chain, Clone: 1C1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433540025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14335403

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Human
Format Affinity Purified
Immunogen The immunogen for this antibody was CD131.
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Conjugate Unlabeled
Antigen Common beta Chain
Formulation Phosphate buffer pH 7.2
Isotype IgG1, κ
Gene ID (Entrez) 1439
Purification Method Affinity Purified
Clone 1C1