Please login to your online account to display your discounted pricing

Mouse anti-Common beta Chain, Clone: 1C1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14335401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14335401

  • / Each
 

Description & Specifications

Specifications

Antigen Common beta Chain
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Cross Reactivity Human
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Mouse
Isotype IgG1, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1439
Clone 1C1
Conjugate Unlabeled
Immunogen The immunogen for this antibody was CD131.
Primary or Secondary Primary
Concentration 0.2mg/mL