Please login to your online account to display your discounted pricing

Mouse anti-Coactosin-like Protein 1/CotL1, Clone: 1D6, Novus Biologicals™

$254.34 - $311.17

Specifications

Antigen Coactosin-like Protein 1/CotL1
Clone 1D6
Concentration 1mg/mL
Immunogen Recombinant human COTL1 (1-142aa) purified from E. coli
Gene ID (Entrez) 23406
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23120301

 
novus biologicals canada
NBP23120301ML
0.1mL Each for $311.17

NBP23120305

 
novus biologicals canada
NBP231203005ML
0.05mL Each for $254.34
Description & Specifications

Specifications

Antigen Coactosin-like Protein 1/CotL1
Clone 1D6
Concentration 1mg/mL
Immunogen Recombinant human COTL1 (1-142aa) purified from E. coli
Gene ID (Entrez) 23406
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blot