Please login to your online account to display your discounted pricing

Mouse anti-CKAP1, Clone: 1F6, Novus Biologicals™

$254.34 - $310.66

Specifications

Antigen CKAP1
Clone 1F6
Concentration 1mg/mL
Immunogen Recombinant human TBCB (1-244aa) purified from E. coli
Gene Accession No. NP_001272
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23120701

 
novus biologicals canada
NBP23120701ML
0.1mL Each for $310.66

NBP23120705

 
novus biologicals canada
NBP231207005ML
0.05mL Each for $254.34
Description & Specifications

Specifications

Antigen CKAP1
Clone 1F6
Concentration 1mg/mL
Immunogen Recombinant human TBCB (1-244aa) purified from E. coli
Gene Accession No. NP_001272
Gene ID (Entrez) 1155
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blotting