Please login to your online account to display your discounted pricing

Mouse anti-CKAP1, Clone: 1F6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231207005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23120705

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen CKAP1
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone 1F6
Applications ELISA
Applications Western Blotting
Formulation Liquid. Supplied in Phosphate-Buffered Saline (pH 7.4) containing 10% Glycerol
Immunogen Recombinant human TBCB (1-244aa) purified from E. coli
Primary or Secondary Primary
Concentration 1mg/mL
Test Specificity Anti-human TBCB mAb, clone AT1F6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human TBCB protein.
Reconstitution Protein A purified
Quantity 0.05mL
Format Purified
Isotype IgG2b, κ
Gene ID (Entrez) 1155
Gene Accession No. NP_001272
Conjugate Unlabeled
Purification Method Purified