Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM585, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Chromogranin A
Clone SPM585
Concentration 0.2mg/mL
Immunogen Human pheochromocytoma
Gene Accession No. P10645
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23295901

 
novus biologicals canada
NBP23295901ML
0.1mL Each for $139.00

NBP23295910

 
novus biologicals canada
NBP23295910ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen Chromogranin A
Clone SPM585
Concentration 0.2mg/mL
Immunogen Human pheochromocytoma
Gene Accession No. P10645
Gene ID (Entrez) 1113
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting