Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM585, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23295901ML

 View all options for this product


For Research Use Only

Catalog No. NBP23295901


Description & Specifications


Antigen Chromogranin A
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM585
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10645
Host Species Mouse
Immunogen Human pheochromocytoma
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Test Specificity Does not react to Mouse and Rat.
Reconstitution Protein A purified