Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM585, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23295910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23295910

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 1113
Concentration 0.2mg/mL
Antigen Chromogranin A
Gene Accession No. P10645
Host Species Mouse
Immunogen Human pheochromocytoma
Isotype IgG1, κ
Purification Method Purified
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone SPM585
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Formulation PBS with 0.05% BSA.
Test Specificity Does not react to Mouse and Rat.