Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM585, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481502

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen Chromogranin A
Immunogen Human pheochromocytoma
Regulatory Status RUO
Format Purified
Conjugate Unlabeled
Clone SPM585
Quantity 0.2mg
Reconstitution Protein A purified
Primary or Secondary Primary
Concentration 1mg/mL
Gene Accession No. P10645
Isotype IgG1, κ
Cross Reactivity Human
Cross Reactivity Primate
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Test Specificity Does not react to Mouse or Rat.
Gene ID (Entrez) 1113
Host Species Mouse