Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM585, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23481501MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23481501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P10645
Immunogen Human pheochromocytoma
Purification Method Purified
Regulatory Status RUO
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled
Antigen Chromogranin A
Host Species Mouse
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Clone SPM585