Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM553, Azide Free, Novus Biologicals™

$527.00 - $2,910.00

Specifications

Antigen Chromogranin A
Clone SPM553
Concentration 1mg/mL
Immunogen Recombinant human chromogranin A protein
Gene Accession No. P10645
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23479501

 
novus biologicals canada
NBP23479501MG
0.1mg Each for $2,910.00

NBP23479502

 
novus biologicals canada
NBP23479502MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Chromogranin A
Clone SPM553
Concentration 1mg/mL
Immunogen Recombinant human chromogranin A protein
Gene Accession No. P10645
Gene ID (Entrez) 1113
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot