Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: SPM339, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23295701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23295701

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human pheochromocytoma
Purification Method Purified
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Test Specificity Does not react to Guinea Pig, Rabbit or Sheep.
Clone SPM339
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Antigen Chromogranin A
Gene Accession No. P10645
Host Species Mouse
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified