Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: PHE5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470902

  • / Each
 

Description & Specifications

Specifications

Antigen Chromogranin A
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P10645
Host Species Mouse
Immunogen Human pheochromocytoma
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Test Specificity Does not react to Mouse or Rat.
Clone PHE5
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Reconstitution Protein A purified
Concentration 1mg/mL