Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: PHE5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23470901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23470901

  • / Each
 

Description & Specifications

Specifications

Antigen Chromogranin A
Gene Accession No. P10645
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human pheochromocytoma
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 1113
Test Specificity Does not react to Mouse or Rat.
Reconstitution Protein A purified
Clone PHE5
Concentration 1mg/mL
Conjugate Unlabeled
Purification Method Purified
Regulatory Status RUO