Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10 + PHE5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423901

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Chromogranin A
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P10645
Host Species Mouse
Immunogen Human pheochromocytoma (LK2H10 & PHE5)
Isotype IgG1, κ
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1113
Reconstitution Protein A purified
Clone LK2H10 + PHE5
Purification Method Purified
Quantity 0.1mL
Concentration 0.2mg/mL