Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10 + PHE5 + CGA414, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23424005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23424005

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Gene Accession No. P10645
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Conjugate Unlabeled
Antigen Chromogranin A
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 1113
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Isotype IgG1
Quantity 0.5mL
Clone LK2H10 + PHE5 + CGA414
Immunogen Human pheochromocytoma (LK2H10 & PHE5) and recombinant human chromogranin A protein (CGA414)
Purification Method Purified
Concentration 0.2mg/mL
Regulatory Status RUO