Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10 + PHE5 + CGA414, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467401

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Immunogen Human pheochromocytoma (LK2H10 & PHE5) and recombinant human chromogranin A protein (CGA414)
Conjugate Unlabeled
Gene Accession No. P10645
Primary or Secondary Primary
Host Species Mouse
Gene ID (Entrez) 1113
Isotype IgG1
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Quantity 0.1mg
Purification Method Purified
Clone LK2H10 + PHE5 + CGA414
Antigen Chromogranin A
Concentration 1mg/mL
Reconstitution Protein A purified