Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10 + PHE5, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Chromogranin A
Clone LK2H10 + PHE5
Concentration 1mg/mL
Immunogen Human pheochromocytoma (LK2H10 & PHE5)
Gene Accession No. P10645
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23467301

 
novus biologicals canada
NBP23467301MG
0.1mg Each for $384.00

NBP23467302

 
novus biologicals canada
NBP23467302MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Chromogranin A
Clone LK2H10 + PHE5
Concentration 1mg/mL
Immunogen Human pheochromocytoma (LK2H10 & PHE5)
Gene Accession No. P10645
Gene ID (Entrez) 1113
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting