Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10 + PHE5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Immunogen Human pheochromocytoma (LK2H10 & PHE5)
Regulatory Status RUO
Format Purified
Host Species Mouse
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.1mg
Gene Accession No. P10645
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Clone LK2H10 + PHE5
Antigen Chromogranin A
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Reconstitution Protein A purified
Concentration 1mg/mL
Gene ID (Entrez) 1113