Please login to your online account to display your discounted pricing

Mouse anti-Chromogranin A, Clone: LK2H10, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Conjugate Unlabeled
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone LK2H10
Primary or Secondary Primary
Antigen Chromogranin A
Gene Accession No. P10645
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1, κ
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Test Specificity Does not react to Guinea Pig, Rabbit or Sheep.
Immunogen Human pheochromocytoma
Purification Method Purified
Format Purified
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Gene ID (Entrez) 1113