Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: SPM551, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479302

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Antigen CEACAM-5/CD66e
Format Purified
Gene Accession No. P06731
Immunogen Human recombinant CEA protein
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 1048
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family.
Reconstitution Protein A purified
Clone SPM551
Concentration 1mg/mL