Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: SPM551, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23479301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23479301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Primate
Immunogen Human recombinant CEA protein
Isotype IgG1, κ
Antigen CEACAM-5/CD66e
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P06731
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Conjugate Unlabeled
Reconstitution Protein A purified
Host Species Mouse
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family.
Format Purified
Gene ID (Entrez) 1048
Purification Method Purified
Clone SPM551
Concentration 1mg/mL
Regulatory Status RUO