Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: SPM541, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478701

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human colon carcinoma extract
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Host Species Mouse
Primary or Secondary Primary
Gene Accession No. P06731
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene ID (Entrez) 1048
Isotype IgG1, κ
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family.
Reconstitution Protein A purified
Antigen CEACAM-5/CD66e
Quantity 0.1mg
Regulatory Status RUO
Clone SPM541
Concentration 1mg/mL
Format Purified