Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: COL-1 + CEA31 + C66/261, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CEACAM-5/CD66e
Clone COL-1 + CEA31 + C66/261
Concentration 0.2mg/mL
Immunogen Human colon carcinoma extract (COL-1 & CEA31); CEA recombinant protein (C66/261)
Gene Accession No. P06731
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23423301

 
novus biologicals canada
NBP23423301ML
0.1mL Each for $139.00

NBP23423305

 
novus biologicals canada
NBP23423305ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CEACAM-5/CD66e
Clone COL-1 + CEA31 + C66/261
Concentration 0.2mg/mL
Immunogen Human colon carcinoma extract (COL-1 & CEA31); CEA recombinant protein (C66/261)
Gene Accession No. P06731
Gene ID (Entrez) 1048
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting