Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: COL-1 + CEA31 + C66/261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423305

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Immunogen Human colon carcinoma extract (COL-1 & CEA31); CEA recombinant protein (C66/261)
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Antigen CEACAM-5/CD66e
Gene Accession No. P06731
Host Species Mouse
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 1048
Reconstitution Protein A purified
Clone COL-1 + CEA31 + C66/261
Conjugate Unlabeled