Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: COL-1 + CEA31 + C66/261, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23423301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23423301

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 1048
Format Purified
Regulatory Status RUO
Conjugate Unlabeled
Antigen CEACAM-5/CD66e
Gene Accession No. P06731
Host Species Mouse
Immunogen Human colon carcinoma extract (COL-1 & CEA31); CEA recombinant protein (C66/261)
Isotype IgG1
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Clone COL-1 + CEA31 + C66/261
Concentration 0.2mg/mL
Cross Reactivity Human
Primary or Secondary Primary