Please login to your online account to display your discounted pricing

Mouse anti-CEACAM-5/CD66e, Clone: COL-1 + CEA31 + C66/261, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459701

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1
Purification Method Purified
Gene ID (Entrez) 1048
Reconstitution Protein A purified
Antigen CEACAM-5/CD66e
Gene Accession No. P06731
Host Species Mouse
Immunogen Human colon carcinoma extract (COL-1 & CEA31); CEA recombinant protein (C66/261)
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Cross Reactivity Human
Primary or Secondary Primary
Clone COL-1 + CEA31 + C66/261
Conjugate Unlabeled