Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: SPM551, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen CEA
Clone SPM551
Concentration 0.2mg/mL
Immunogen Human recombinant CEA protein
Gene Accession No. P06731
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23301101

 
novus biologicals canada
NBP23301101ML
0.1mL Each for $139.00

NBP23301110

 
novus biologicals canada
NBP23301110ML
1.0mL Each for $429.00
Description & Specifications

Specifications

Antigen CEA
Clone SPM551
Concentration 0.2mg/mL
Immunogen Human recombinant CEA protein
Gene Accession No. P06731
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting