Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: SPM541, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23301301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23301301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Primate
Antigen CEA
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P06731
Immunogen Human colon carcinoma extract
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Reconstitution Protein A purified
Clone SPM541
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse
Monoclonal or Polyclonal Monoclonal