Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: COL-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23300910ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23300910

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen CEA
Format Purified
Formulation PBS with 0.05% BSA.
Quantity 1.0mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone COL-1
Conjugate Unlabeled
Gene Accession No. P06731
Host Species Mouse
Immunogen Human colon carcinoma extract
Isotype IgG2a, κ
Purification Method Purified
Primary or Secondary Primary
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues.
Reconstitution Protein A purified
Concentration 0.2mg/mL