Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: COL-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23300901ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23300901

  • / Each
 

Description & Specifications

Specifications

Antigen CEA
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Cross Reactivity Human
Gene Accession No. P06731
Host Species Mouse
Immunogen Human colon carcinoma extract
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Formulation PBS with 0.05% BSA.
Purification Method Purified
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues.
Clone COL-1
Conjugate Unlabeled