Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: COL-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23485101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23485101

  • / Each
 

Description & Specifications

Specifications

Antigen CEA
Format Purified
Formulation PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06731
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Concentration 1.0mg/mL
Host Species Mouse
Immunogen Human colon carcinoma extract
Isotype IgG2a, κ
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody recognizes proteins of 80-200kDa, identified as different members of CEA family. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues.
Reconstitution Protein A purified
Clone COL-1
Conjugate Unlabeled
Cross Reactivity Human