Please login to your online account to display your discounted pricing

Mouse anti-CEA, Clone: CEA31, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23459401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23459401

  • / Each
 

Description & Specifications

Specifications

Antigen CEA
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human colon carcinoma extract
Isotype IgG1, κ
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Primate
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06731
Host Species Mouse
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone CEA31
Conjugate Unlabeled
Reconstitution Protein A purified
Concentration 1mg/mL