Please login to your online account to display your discounted pricing

Mouse anti-CDw60, Clone: UM4D4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446605

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Immunogen Human rheumatoid synovial T cell line (ST-1)
Conjugate Unlabeled
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Isotype IgM
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Antigen CDw60
Purification Method Concentrated
Clone UM4D4