Please login to your online account to display your discounted pricing

Mouse anti-CDw60, Clone: UM4D4, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23446601ML

 View all options for this product


For Research Use Only

Catalog No. NBP23446601


Description & Specifications


Antigen CDw60
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Clone UM4D4
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Immunogen Human rheumatoid synovial T cell line (ST-1)
Isotype IgM
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal