Please login to your online account to display your discounted pricing

Mouse anti-Cdc20, Clone: AR12, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Cdc20
Clone AR12
Concentration 1mg/mL
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Gene Accession No. Q12834
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23455201

 
novus biologicals canada
NBP23455201MG
0.1mg Each for $384.00

NBP23455202

 
novus biologicals canada
NBP23455202MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Cdc20
Clone AR12
Concentration 1mg/mL
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Gene Accession No. Q12834
Gene ID (Entrez) 991
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot