Please login to your online account to display your discounted pricing

Mouse anti-Cdc20, Clone: AR12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23455202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23455202

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 991
Antigen Cdc20
Gene Accession No. Q12834
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone AR12
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human