Please login to your online account to display your discounted pricing

Mouse anti-Cdc20, Clone: AR12, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23455201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23455201

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Antigen Cdc20
Cross Reactivity Human
Primary or Secondary Primary
Gene Accession No. Q12834
Conjugate Unlabeled
Purification Method Purified
Clone AR12
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Immunogen Urea-denatured His6 Cdc20 human recombinant protein
Regulatory Status RUO
Gene ID (Entrez) 991
Concentration 1mg/mL
Reconstitution Protein A purified
Isotype IgG1, κ