Please login to your online account to display your discounted pricing

Mouse anti-CDC2/CDK1, Clone: POH-1, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CDC2/CDK1
Clone POH-1
Concentration 0.2mg/mL
Immunogen Purified recombinant human p34cdc2 fusion protein
Gene Accession No. P06493
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23433101

 
novus biologicals canada
NBP23433101ML
0.1mL Each for $139.00

NBP23433105

 
novus biologicals canada
NBP23433105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CDC2/CDK1
Clone POH-1
Concentration 0.2mg/mL
Immunogen Purified recombinant human p34cdc2 fusion protein
Gene Accession No. P06493
Gene ID (Entrez) 983
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mink
Cross Reactivity Primate
Isotype IgG2a, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting