Please login to your online account to display your discounted pricing

Mouse anti-CDC2/CDK1, Clone: POH-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23433105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23433105

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mink
Cross Reactivity Primate
Format Purified
Gene Accession No. P06493
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified recombinant human p34cdc2 fusion protein
Isotype IgG2a, κ
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 983
Clone POH-1
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a 34kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1/p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1/p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation. Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative). Does not react with S. pombe, S. cerevisiae.
Reconstitution Protein A purified
Concentration 0.2mg/mL
Antigen CDC2/CDK1