missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-CDC2/CDK1, Clone: POH-1, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen CDC2/CDK1
Clone POH-1
Concentration 1mg/mL
Immunogen Purified recombinant human p34cdc2 fusion protein
Gene Accession No. P06493
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23471101 Novus Biologicals Canada
NBP23471101MG
0.1mg Each for $384.00
NBP23471102 Novus Biologicals Canada
NBP23471102MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

CDC2/CDK1
1mg/mL
P06493
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
POH-1
Purified recombinant human p34cdc2 fusion protein
983
Monoclonal
Store at -20 to -80°C. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot