Please login to your online account to display your discounted pricing

Mouse anti-CDC2/CDK1, Clone: POH-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471101

  • / Each
 

Description & Specifications

Specifications

Antigen CDC2/CDK1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone POH-1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P06493
Host Species Murine
Immunogen Purified recombinant human p34cdc2 fusion protein
Isotype IgG2a, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 983
Test Specificity Recognizes a 34kDa protein, which is identified as cyclin dependent kinase 1 (cdk1) or p34cdc2 protein kinase. cdk1/p34cdc2 plays a crucial role during cell division and is most active during mitosis. It is predominantly localized in the nucleus. It is a serine/threonine kinase, which is activated by cyclin, presumably by de-phosphorylation of tyrosine residues. Activated cdk1/p34cdc2 performs specific functions during mitosis, including nuclear envelope breakdown and chromosome condensation. Drosophila (Negative), Mouse (Negative), Rat (Negative), Xenopus (Negative). Does not react with S. pombe, S. cerevisiae.
Reconstitution Protein A purified
Target Species Bovine
Target Species Human
Target Species Mink
Target Species Primate