Please login to your online account to display your discounted pricing

Mouse anti-CD99, Clone: SPM586, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen CD99
Clone SPM586
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Gene Accession No. P14209
Gene ID (Entrez) 4267
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445701

 
novus biologicals canada
NBP23445701ML
0.1mL Each for $139.00

NBP23445705

 
novus biologicals canada
NBP23445705ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen CD99
Clone SPM586
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Gene Accession No. P14209
Gene ID (Entrez) 4267
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Rat
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)