Please login to your online account to display your discounted pricing

Mouse anti-CD99, Clone: SPM586, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23445701ML

 View all options for this product


For Research Use Only

Catalog No. NBP23445701


Description & Specifications


Antigen CD99
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM586
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P14209
Host Species Mouse
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4267
Test Specificity Recognizes a sialoglycoprotein of 27-32kDa, identified as CD99, or MIC2 gene product, or E2 antigen. This antigen is expressed on the cell membrane of some lymphocytes, cortical thymocytes, and granulosa cells of the ovary.