Please login to your online account to display your discounted pricing

Mouse anti-CD99, Clone: HO36-1.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446505

  • / Each
 

Description & Specifications

Specifications

Antigen CD99
Format Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Rat
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P14209
Host Species Mouse
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Isotype IgM, κ
Purification Method Concentrated
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4267
Conjugate Unlabeled
Quantity 0.5mL
Clone HO36-1.1