Please login to your online account to display your discounted pricing

Mouse anti-CD99, Clone: HO36-1.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446501

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Mouse
Gene Accession No. P14209
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL
Format Concentrated
Immunogen Purified E-rosette forming cells from human peripheral blood lymphocytes
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 4267
Conjugate Unlabeled
Antigen CD99
Clone HO36-1.1
Isotype IgM, κ
Regulatory Status RUO
Purification Method Concentrated
Cross Reactivity Human
Cross Reactivity Rat
Formulation Culture Medium. Bioreactor concentrate.