Please login to your online account to display your discounted pricing

Mouse anti-CD99, Clone: 3B2/TA8, PE, Novus Biologicals™

$296.00 - $296.00

Specifications

Antigen CD99
Clone 3B2/TA8
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD99.
Gene Accession No. P14209
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143831

 
novus biologicals canada
NBP143831100TESTS
100 Tests Each for $296.00
Description & Specifications

Specifications

Antigen CD99
Clone 3B2/TA8
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD99.
Gene Accession No. P14209
Gene ID (Entrez) 4267
Formulation Aqueous buffer (contains stabilizer if necessary)
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry